Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethylimidazole ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

E0233-25G E0233-500G
TCE0233-025GEA 32 USD InStock
TCE0233-025G TCE0233-500G
2-Ethylimidazole ≥98.0% (by GC, titration analysis)
2-Ethylimidazole
CAS Number: 1072-62-4
MDL Number: MFCD00005192
Molecular Formula: C5H8N2
Molecular Weight: 96.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 83
Flash Point (°C): 159


Formula: C₅H₈N₂
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00005192
CAS Number: 1072-62-4
EINECS: 214-011-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a367b2e4152d4ad14aaef6802146a98d98abf447&vid=a367b2e4152d4ad14aaef6802146a98d98abf447&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty