Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorophenylhydrazinium chloride ≥97.0%

Supplier: Decon Laboratories Ltd

F0368-5G F0368-1G
TCF0368-005GEA 64 USD InStock
TCF0368-005G TCF0368-1G
2-Fluorophenylhydrazinium chloride ≥97.0%
2-Fluorophenylhydrazinium chloride
CAS Number: 2924-15-4
MDL Number: MFCD00012927
Molecular Formula: C6H7FN2
Molecular Weight: 162.59
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Formula: C₆H₈ClFN₂
MW: 162.59 g/mol
MDL Number: MFCD00012927
CAS Number: 2924-15-4
EINECS: 220-885-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=051f19a5dd86671f060a04b47a9316821bdbdbc1&vid=051f19a5dd86671f060a04b47a9316821bdbdbc1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty