Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Heptanal ≥95.0%

Supplier: Decon Laboratories Ltd
Danger

H0025-500ML H0025-25ML
TCH0025-500MLEA 136 USD InStock
TCH0025-500ML TCH0025-25ML
Heptanal ≥95.0%
Heptanal
CAS Number: 111-71-7
MDL Number: MFCD00007028
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 34
Specific Gravity (20/20): 0.82

Other Product Names / Synonyms: Enanthal, Enanthaldehyde, Heptaldehyde, Heptyl Aldehyde


Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 153
Melting Pt: -43
Density: 0.82 (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00007028
CAS Number: 111-71-7
EINECS: 203-898-4
UN: 3056
ADR: 3 III
Merck Index: 13,04677
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad313f6259f1910211c4a748a88d1d35fea4c1f1&vid=ad313f6259f1910211c4a748a88d1d35fea4c1f1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty