Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Heptanone ≥97.0%

Supplier: Decon Laboratories Ltd
Danger

H0039-500ML H0039-25ML
TCH0039-500MLEA 211 USD InStock
TCH0039-500ML TCH0039-25ML
4-Heptanone ≥97.0%
4-Heptanone
CAS Number: 123-19-3
MDL Number: MFCD00009403
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 143
Flash Point (°C): 37
Specific Gravity (20/20): 0.82

Other Product Names / Synonyms: Butyrone, Dipropyl Ketone


Formula: C₇H₁₄O
Boiling Pt: 143 °C
Melting Pt: -34 °C
Density: 0.82 (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00009403
CAS Number: 123-19-3
EINECS: 204-608-9
UN: 2710
ADR: 3,III
Merck Index: 13,03379
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97591482d39f80dbdbd4ead24cf735cc1299dd59&vid=97591482d39f80dbdbd4ead24cf735cc1299dd59&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty