Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexamethyldisiloxane, 0.65 cSt (25°C) ≥98.0%

Supplier: Decon Laboratories Ltd

H0091-500ML H0091-25ML
TCH0091-500MLEA 63.52 USD InStock
TCH0091-500ML TCH0091-25ML
Hexamethyldisiloxane, 0.65 cSt (25°C) ≥98.0%
Hexamethyldisiloxane
CAS Number: 107-46-0
MDL Number: MFCD00008265
Molecular Formula: C6H18OSi2
Molecular Weight: 162.38
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): -2
Specific Gravity (20/20): 0.77


Formula: C₆H₁₈OSi₂
MDL Number: MFCD00008265
CAS Number: 107-46-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed543d0ab0f5b3cb3a35f470960e65a7f4aaf543&vid=ed543d0ab0f5b3cb3a35f470960e65a7f4aaf543&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty