Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexanoyl chloride ≥98.0%

Supplier: Decon Laboratories Ltd
Danger

H0117-500G H0117-25G H0117-100G
TCH0117-500GEA 138 USD InStock
TCH0117-500G TCH0117-25G TCH0117-100G
Hexanoyl chloride ≥98.0%
Hexanoyl chloride
CAS Number: 142-61-0
MDL Number: MFCD00000760
Molecular Formula: C6H11ClO
Molecular Weight: 134.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 50
Specific Gravity (20/20): 0.98


Formula: C₆H₁₁ClO
MW: 134.61 g/mol
Boiling Pt: 152
Melting Pt: -87
Density: 0.98 (20 °C)
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00000760
CAS Number: 142-61-0
EINECS: 205-549-1
UN: 2920
ADR: 8 II
Merck Index: 13,01769
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c836feafee35e996378e85f12f8b299f1d7dd77&vid=0c836feafee35e996378e85f12f8b299f1d7dd77&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty