Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-11-Hexadecenal ≥95.0%

Supplier: Decon Laboratories Ltd
Warning

H0679-1G H0679-10G
TCH0679-001GEA 98 USD InStock
TCH0679-001G TCH0679-010G
cis-11-Hexadecenal ≥95.0%
cis-11-Hexadecenal
CAS Number: 53939-28-9
MDL Number: MFCD00010124
Molecular Formula: C16H30O
Molecular Weight: 238.42
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 61
Specific Gravity (20/20): 0.85
Storage Temperature: 0-10°C

Other Product Names / Synonyms: cis-11-Hexadecenyl Aldehyde


Formula: C₁₆H₃₀O
Boiling Pt: 132 °C
Density: 0.85 (20 °C)
Flash Pt: 61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010124
CAS Number: 53939-28-9
EINECS: 258-876-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c08cbadb51986aaf1e03397c04516f6ba378d0c&vid=8c08cbadb51986aaf1e03397c04516f6ba378d0c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty