Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole ≥99.0%

Supplier: Decon Laboratories Ltd
Danger

I0021-500G I0021-25G
TCI0021-500GEA 240 USD InStock
TCI0021-500G TCI0021-25G
Indole ≥99.0%
Indole
CAS Number: 120-72-9
MDL Number: MFCD00005607
Molecular Formula: C8H7N
Molecular Weight: 117.15
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 253
Melting point (°C): 53
Flash Point (°C): 122


Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 253 °C
Melting Pt: 53 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 122 °C
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1 III
Merck Index: 13,04985
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d0985c6da19418f498fb4b4e83f5103cc828dbe&vid=5d0985c6da19418f498fb4b4e83f5103cc828dbe&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty