Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopentyl nitrite (mixed isomers ) ≥95.0%

Supplier: Decon Laboratories Ltd
Danger

I0089-500ML I0089-25ML
TCI0089-500MLEA 171.42 USD InStock
TCI0089-500ML TCI0089-25ML
Isopentyl nitrite (mixed isomers ) ≥95.0%
Isopentyl nitrite (mixed isomers )
CAS Number: 110-46-3
MDL Number: MFCD00002057
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 3
Specific Gravity (20/20): 0.88
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Reagents for the Generation of Arynes (Aryne Precursors), Bioscience, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: Isopentyl Nitrite, 3-Methylbutyl Nitrite


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 99 °C
Density: 0.88 (20 °C)
Flash Pt: 3 °C
MDL Number: MFCD00002057
CAS Number: 110-46-3
EINECS: 203-770-8
UN: 1113
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d13dae4c60bc48cd338211fdc064208fc8fe529c&vid=d13dae4c60bc48cd338211fdc064208fc8fe529c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty