Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Isopropylbenzaldehyde ≥97.0%

Supplier: Decon Laboratories Ltd
Warning

I0168-25G I0168-500G
TCI0168-025GEA 29.49 USD InStock
TCI0168-025G TCI0168-500G
4-Isopropylbenzaldehyde ≥97.0%
4-Isopropylbenzaldehyde
CAS Number: 122-03-2
MDL Number: MFCD00006953
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 108
Specific Gravity (20/20): 0.98

Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: Cuminic Aldehyde, 4-Isopropylbenzaldehyde


Formula: C₁₀H₁₂O
Boiling Pt: 240 °C
Density: 0.98 (20 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00006953
CAS Number: 122-03-2
EINECS: 204-516-9
Merck Index: 13,02648
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=947ed26f676f89be978d9dee7e67772815c07aff&vid=947ed26f676f89be978d9dee7e67772815c07aff&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty