I0200-500ML
I0200-25ML
I0200-100ML
TCI0200-500MLEA
216
USD
InStock
TCI0200-500ML
TCI0200-25ML
TCI0200-100ML
Isovaleryl chloride ≥99.0%
Isovaleryl chloride
CAS Number: 108-12-3
MDL Number: MFCD00000738
Molecular Formula: C5H9ClO
Molecular Weight: 120.58
Purity/Analysis Method: >99.0% (T)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 18
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: Isovaleroyl Chloride