L0226-25G
L0226-500G
TCL0226-25GEA
35.39
USD
InStock
TCL0226-25G
TCL0226-500G
Lactic acid ≥85.0%
Lactic acid
CAS Number: 50-21-5
MDL Number: MFCD00004520
Molecular Formula: C3H6O3
Molecular Weight: 90.08
Purity/Analysis Method: >85.0% (T)
Form: Clear Liquid
Boiling point (°C): 122
Melting point (°C): 17
Flash Point (°C): 110
Specific Gravity (20/20): 1.21
Other Product Names / Synonyms: 2-Hydroxypropionic Acid