Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Maleic anhydride ≥99.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd
Danger

M0005-25G M0005-500G
TCM0005-25GEA 23.41 USD InStock
TCM0005-25G TCM0005-500G
Maleic anhydride ≥99.0% (by GC, titration analysis)
Maleic anhydride
CAS Number: 108-31-6
MDL Number: MFCD00005518
Molecular Formula: C4H2O3
Molecular Weight: 98.06
Purity/Analysis Method: >99.0% (GC,T)
Form: Pellet
Boiling point (°C): 199
Melting point (°C): 54
Flash Point (°C): 102


Formula: C₄H₂O₃
Boiling Pt: 199 °C
Melting Pt: 54 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00005518
CAS Number: 108-31-6
EINECS: 203-571-6
UN: 2215
ADR: 8,III
Merck Index: 13,05727
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9abd3e41098c808e06c986c5d87d1ba8b2ac52fd&vid=9abd3e41098c808e06c986c5d87d1ba8b2ac52fd&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty