M0173-500ML
M0173-25ML
TCM0173-500MLEA
92.21
USD
InStock
TCM0173-500ML
TCM0173-25ML
3-Methylbutanone ≥99.0%
3-Methylbutanone
CAS Number: 563-80-4
MDL Number: MFCD00008919
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Melting point (°C): -92
Flash Point (°C): 1
Specific Gravity (20/20): 0.80
Other Product Names / Synonyms: Isopropyl Methyl Ketone