Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Mercaptobenzothiazole ≥99.0%

Supplier: Decon Laboratories Ltd
Warning

M0247-500G M0247-25G
TCM0247-500GEA 155 USD InStock
TCM0247-500G TCM0247-25G
2-Mercaptobenzothiazole ≥99.0%
2-Mercaptobenzothiazole
CAS Number: 149-30-4
MDL Number: MFCD00005781
Molecular Formula: C7H5NS2
Molecular Weight: 167.24
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 180
Flash Point (°C): 200
Lambda max.: 325 nm (MeOH)


Formula: C₇H₅NS₂
MW: 167.26 g/mol
Boiling Pt: 302
Melting Pt: 180 °C
Density: 1.42 g/cm³ (20 °C)
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00005781
CAS Number: 149-30-4
EINECS: 205-736-8
UN: 3077
ADR: 9 III
Merck Index: 13,05893
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cc4f6b9b8585329256e9824df3020684633ade2&vid=5cc4f6b9b8585329256e9824df3020684633ade2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty