Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl myristate ≥98.0%

Supplier: Decon Laboratories Ltd

M0482-25ML M0482-500ML
TCM0482-025MLEA 17.9 USD InStock
TCM0482-025ML TCM0482-500ML
Methyl myristate ≥98.0%
Methyl myristate
CAS Number: 124-10-7
MDL Number: MFCD00008983
Molecular Formula: C15H30O2
Molecular Weight: 242.40
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Flash Point (°C): 157
Freezing point (°C): 18
Specific Gravity (20/20): 0.87

Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Methyl Tetradecanoate, Myristic Acid Methyl Ester


Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Boiling Pt: 323
Melting Pt: 18
Density: 0.87 (20 °C)
Flash Pt: 157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008983
CAS Number: 124-10-7
EINECS: 204-680-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27ca0087c2121f54002e6007a5ecf1697236682d&vid=27ca0087c2121f54002e6007a5ecf1697236682d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty