Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Methoxyphenyl)propionic acid ≥98.0%

Supplier: Decon Laboratories Ltd

M1032-25G M1032-5G
TCM1032-025GEA 191 USD InStock
TCM1032-025G TCM1032-005G
3-(4-Methoxyphenyl)propionic acid ≥98.0%
3-(4-Methoxyphenyl)propionic acid
CAS Number: 1929-29-9
MDL Number: MFCD00002777
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 194
Melting point (°C): 104


Formula: C₁₀H₁₂O₃
Boiling Pt: 194 °C
Melting Pt: 104 °C
MDL Number: MFCD00002777
CAS Number: 1929-29-9
EINECS: 217-678-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e237cd9d313c6b4ded1a555534c071f1b9827fd9&vid=e237cd9d313c6b4ded1a555534c071f1b9827fd9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty