Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Naphthoquinone ≥98.0%

Supplier: Decon Laboratories Ltd
Danger

N0040-500G N0040-25G N0040-100G
TCN0040-500GEA 197 USD InStock
TCN0040-500G TCN0040-25G TCN0040-100G
1,4-Naphthoquinone ≥98.0%
1,4-Naphthoquinone
CAS Number: 130-15-4
MDL Number: MFCD00001676
Molecular Formula: C10H6O2
Molecular Weight: 158.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 125


Formula: C₁₀H₆O₂
MW: 158.16 g/mol
Boiling Pt: 298
Melting Pt: 124
Density: 1.422 g/cm³ (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00001676
CAS Number: 130-15-4
EINECS: 204-977-6
UN: 2811
ADR: 6.1 I
Merck Index: 12,06482
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3368db37bc7a7041b81f7b28843d42d684ce0af&vid=b3368db37bc7a7041b81f7b28843d42d684ce0af&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty