Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nitrilotriacetic acid ≥98.0%

Supplier: Decon Laboratories Ltd
Warning

N0098-25G N0098-500G
TCN0098-025GEA 29.94 USD InStock
TCN0098-025G TCN0098-500G
Nitrilotriacetic acid ≥98.0%
Nitrilotriacetic acid
CAS Number: 139-13-9
MDL Number: MFCD00004287
Molecular Formula: C6H9NO6
Molecular Weight: 191.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 245
Flash Point (°C): 100

Other Product Names / Synonyms: Triglycollamic Acid, NTA


Formula: C₆H₉NO₆
Boiling Pt: 498 °C (1013 hPa)
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004287
CAS Number: 139-13-9
EINECS: 205-355-7
UN: 2923
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f625a3fdac71c8aa4cebcf3936badbc4d6ade19c&vid=f625a3fdac71c8aa4cebcf3936badbc4d6ade19c&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty