Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethyl-4-nitrosoaniline ≥98.0%

Supplier: Decon Laboratories Ltd

N0262-500G N0262-25G
TCN0262-500GEA 865 USD InStock
TCN0262-500G TCN0262-25G
N,N-Dimethyl-4-nitrosoaniline ≥98.0%
N,N-Dimethyl-4-nitrosoaniline
CAS Number: 138-89-6
MDL Number: MFCD00002063
Molecular Formula: C8H10N2O
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 85

Other Product Names / Synonyms: 4-Nitroso-N,N-dimethylaniline


Formula: C₈H₁₀N₂O
Boiling Pt: 259 °C (1013 hPa)
Density: 1.145 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002063
CAS Number: 138-89-6
EINECS: 205-343-1
UN: 1369
ADR: 4.2,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c77b2ebc11a969b18508008b6a4b163969a7d497&vid=c77b2ebc11a969b18508008b6a4b163969a7d497&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty