Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl octanoate ≥99.0%

Supplier: Decon Laboratories Ltd

O0033-25ML O0033-500ML
TCO0033-025MLEA 17.3 USD InStock
TCO0033-025ML TCO0033-500ML
Methyl octanoate ≥99.0%
Methyl octanoate
CAS Number: 111-11-5
MDL Number: MFCD00009551
Molecular Formula: C9H18O2
Molecular Weight: 158.24
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 76
Specific Gravity (20/20): 0.88

Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: n-Octanoic Acid Methyl Ester


Formula: C₉H₁₈O₂
Boiling Pt: 193 °C
Melting Pt: -36.7 °C
Density: 0.88 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00009551
CAS Number: 111-11-5
EINECS: 203-835-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10b1f053b5fc350ae6bacc125c0d55a4e7bdcaac&vid=10b1f053b5fc350ae6bacc125c0d55a4e7bdcaac&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty