O0044-500ML
O0044-25ML
TCO0044-500MLEA
87.9
USD
InStock
TCO0044-500ML
TCO0044-25ML
Caprylaldehyde ≥98.0%
Caprylaldehyde
CAS Number: 124-13-0
MDL Number: MFCD00007029
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 52
Specific Gravity (20/20): 0.82
Other Product Names / Synonyms: n-Octanaldehyde, n-Octyl Aldehyde