Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Pentanethiol ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

P0053-500ML P0053-25ML
TCP0053-500MLEA 496 USD InStock
TCP0053-500ML TCP0053-25ML
1-Pentanethiol ≥98.0% (by GC, titration analysis)
1-Pentanethiol
CAS Number: 110-66-7
MDL Number: MFCD00004907
Molecular Formula: C5H12S
Molecular Weight: 104.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 18
Specific Gravity (20/20): 0.84

Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Amyl Mercaptan, Pentyl Mercaptan


Formula: C₅H₁₂S
Boiling Pt: 126 °C
Melting Pt: –76 °C
Density: 0.84 (20 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00004907
CAS Number: 110-66-7
EINECS: 203-789-1
UN: 1111
ADR: 3,II
Merck Index: 12,00653
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c5a7b664098e75a9703a7f6d28ce27ffa626987&vid=7c5a7b664098e75a9703a7f6d28ce27ffa626987&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty