Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazole ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

P0546-25G P0546-250G
TCP0546-025GEA 42 USD InStock
TCP0546-025G TCP0546-250G
1H-Pyrazole ≥98.0% (by GC, titration analysis)
1H-Pyrazole
CAS Number: 288-13-1
MDL Number: MFCD00005234
Molecular Formula: C3H4N2
Molecular Weight: 68.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 188
Melting point (°C): 69

Other Product Names / Synonyms: 1,2-Diazole


Formula: C₃H₄N₂
MDL Number: MFCD00005234
CAS Number: 288-13-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5518f87aca4642fbb409973cab7e2bd2cafbffc&vid=d5518f87aca4642fbb409973cab7e2bd2cafbffc&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty