Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,15-Pentadecanedioic acid ≥97.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

P0909-1G P0909-10G
TCP0909-001GEA 31 USD InStock
TCP0909-001G TCP0909-010G
1,15-Pentadecanedioic acid ≥97.0% (by GC, titration analysis)
1,15-Pentadecanedioic acid
CAS Number: 1460-18-0
MDL Number: MFCD00039534
Molecular Formula: C15H28O4
Molecular Weight: 272.39
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 175
Melting point (°C): 114


Formula: C₁₅H₂₈O₄
MDL Number: MFCD00039534
CAS Number: 1460-18-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0057a72e3b542051a4ffdfedeca60b2c304c7fdb&vid=0057a72e3b542051a4ffdfedeca60b2c304c7fdb&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty