Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenoxybenzoic acid ≥98.0%

Supplier: Decon Laboratories Ltd
Warning

P1253-25G P1253-5G
TCP1253-025GEA 157 USD InStock
TCP1253-025G TCP1253-5G
3-Phenoxybenzoic acid ≥98.0%
3-Phenoxybenzoic acid
CAS Number: 3739-38-6
MDL Number: MFCD00002498
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 147


Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 365
Melting Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00002498
CAS Number: 3739-38-6
EINECS: 223-121-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ec69bebf18844e83efb8c954e1efde285fb128f&vid=1ec69bebf18844e83efb8c954e1efde285fb128f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty