Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Tetradecene ≥90.0%

Supplier: Decon Laboratories Ltd

T0087-25ML T0087-500ML
TCT0087-025MLEA 54.39 USD InStock
TCT0087-025ML TCT0087-500ML
1-Tetradecene ≥90.0%
1-Tetradecene
CAS Number: 1120-36-1
MDL Number: MFCD00008981
Molecular Formula: C14H28
Molecular Weight: 196.38
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 232
Flash Point (°C): 112
Specific Gravity (20/20): 0.77


Formula: C₁₄H₂₈
Boiling Pt: 232 °C
Melting Pt: –13 °C
Density: 0.77 (20 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00008981
CAS Number: 1120-36-1
EINECS: 214-306-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94ff995ac493fa9a6a7325912af86d7ac1eb2092&vid=94ff995ac493fa9a6a7325912af86d7ac1eb2092&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >90.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty