Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,3,3-Tetramethylguanidine ≥99.0%

Supplier: Decon Laboratories Ltd
Danger

T0148-25ML T0148-100ML
TCT0148-25MLEA 20 USD InStock
TCT0148-25ML TCT0148-100ML
1,1,3,3-Tetramethylguanidine ≥99.0%
1,1,3,3-Tetramethylguanidine
CAS Number: 80-70-6
MDL Number: MFCD00008323
Molecular Formula: C5H13N3
Molecular Weight: 115.18
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 163
Melting point (°C): -30
Flash Point (°C): 60
Specific Gravity (20/20): 0.92

Other Product Names / Synonyms: TMG


Formula: C₅H₁₃N₃
MW: 115.18 g/mol
Boiling Pt: 163 °C
Melting Pt: –30 °C
Density: 0.92 (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008323
CAS Number: 80-70-6
EINECS: 201-302-7
UN: 2920
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=826b8c08510a79c2ccc425c9719ef2cec4755532&vid=826b8c08510a79c2ccc425c9719ef2cec4755532&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty