T0548-500ML
T0548-25ML
T0548-100ML
TCT0548-500MLEA
96.64
USD
InStock
TCT0548-500ML
TCT0548-25ML
TCT0548-100ML
N,N,N',N'-Tetramethyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
1,3-Bis(dimethylamino)propane
CAS Number: 110-95-2
MDL Number: MFCD00008337
Molecular Formula: C7H18N2
Molecular Weight: 130.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 145
Flash Point (°C): 56
Specific Gravity (20/20): 0.78
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 1,3-Bis(dimethylamino)propane, N,N,N',N'-Tetramethyl-1,3-propanediamine, N,N,N',N'-Tetramethyltrimethylenediamine