Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Titanium(IV) ethanolate

Supplier: Decon Laboratories Ltd

T0967-500ML T0967-25ML
TCT0967-500MLEA 213.76 USD InStock
TCT0967-500ML TCT0967-25ML
Titanium(IV) ethanolate
Titanium(IV) ethanolate
CAS Number: 3087-36-3
MDL Number: MFCD00009071
Molecular Formula: C8H20O4Ti
Molecular Weight: 228.11
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 236
Flash Point (°C): 28
Specific Gravity (20/20): 1.09

Other Product Names / Synonyms: Tetraethyl Titanate, Titanium(IV) Ethoxide


Formula: C₈H₂₀O₄Ti
MW: 228.12 g/mol
Boiling Pt: 150
Density: 1.09 (20 °C)
Flash Pt: 28 °C
MDL Number: MFCD00009071
CAS Number: 3087-36-3
EINECS: 221-410-8
UN: 2924
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=faffe82b73fcdbf04cf64cc7bf692bd5da767c57&vid=faffe82b73fcdbf04cf64cc7bf692bd5da767c57&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty