Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitro-4-(trifluoromethyl)anisole

Supplier: Decon Laboratories Ltd

T1042-5G T1042-25G
TCT1042-005GEA 31 USD InStock
TCT1042-005G TCT1042-025G
2-Nitro-4-(trifluoromethyl)anisole
2-Nitro-4-(trifluoromethyl)anisole
CAS Number: 394-25-2
MDL Number: MFCD00007099
Molecular Formula: C8H6F3NO3
Molecular Weight: 221.14
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 48
Flash Point (°C): 113

Other Product Names / Synonyms: 1-Methoxy-2-nitro-4-(trifluoromethyl)benzene, 4-Trifluoromethyl-2-nitroanisole


Formula: C₈H₆F₃NO₃
Boiling Pt: 90 - 95 °C (0.5 -rr)
Melting Pt: 48 °C
Flash Pt: 113 °C
MDL Number: MFCD00007099
CAS Number: 394-25-2
EINECS: 206-891-4
UN: 1759
ADR: 8,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70652751841d1c2379856d76ec7f4841fdddc604&vid=70652751841d1c2379856d76ec7f4841fdddc604&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty