Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,2-Triphenylacetophenone ≥99.0%

Supplier: Decon Laboratories Ltd

T1207-5G T1207-25G
TCT1207-005GEA 40.19 USD InStock
TCT1207-005G TCT1207-025G
2,2,2-Triphenylacetophenone ≥99.0%
2,2,2-Triphenylacetophenone
CAS Number: 466-37-5
MDL Number: MFCD00004762
Molecular Formula: C26H20O
Molecular Weight: 348.45
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 184
Flash Point (°C): 115

Other Product Names / Synonyms: Benzopinacolone, Phenyl Trityl Ketone


Formula: C₂₆H₂₀O
Boiling Pt: 498 °C (1013 hPa)
Melting Pt: 184 °C
Flash Pt: 115 °C
MDL Number: MFCD00004762
CAS Number: 466-37-5
EINECS: 207-375-1
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty