Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Thiodiacetic acid ≥98.0%

Supplier: Decon Laboratories Ltd
Warning

T1248-500G T1248-25G
TCT1248-500GEA 273 USD InStock
TCT1248-500G TCT1248-25G
2,2'-Thiodiacetic acid ≥98.0%
2,2'-Thiodiacetic acid
CAS Number: 123-93-3
MDL Number: MFCD00004359
Molecular Formula: C4H6O4S
Molecular Weight: 150.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 130

Other Product Names / Synonyms: 2,2'-Thiodiacetic Acid


Formula: C₄H₆O₄S
MW: 150.16 g/mol
Boiling Pt: 168
Melting Pt: 130 °C
Density: 1.661 g/cm³ (20 °C)
MDL Number: MFCD00004359
CAS Number: 123-93-3
EINECS: 204-663-9
UN: 3261
ADR: 8 III
Merck Index: 12,09467
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29434b67af87dc276b35310a5474aa0a3c1a875b&vid=29434b67af87dc276b35310a5474aa0a3c1a875b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty