Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Aminopropyl)trimethoxysilane ≥96.0%

Supplier: Decon Laboratories Ltd
Warning

T1255-500ML T1255-25ML
TCT1255-500MLEA 136 USD InStock
TCT1255-500ML TCT1255-25ML
(3-Aminopropyl)trimethoxysilane ≥96.0%
(3-Aminopropyl)trimethoxysilane
CAS Number: 13822-56-5
MDL Number: MFCD00008206
Molecular Formula: C6H17NO3Si
Molecular Weight: 179.29
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 92
Flash Point (°C): 86
Specific Gravity (20/20): 1.02

Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: 3-(Trimethoxysilyl)propylamine


Formula: H₂NCH₂CH₂CH₂Si(OCH₃)₃
MW: 179.29 g/mol
Boiling Pt: 194
Density: 1.02 (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00008206
CAS Number: 13822-56-5
EINECS: 237-511-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c41d614ba064bf2c6322083519a165138bb684c&vid=8c41d614ba064bf2c6322083519a165138bb684c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty