Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Tetrahydro-2-furoic acid ≥98.0%

Supplier: Decon Laboratories Ltd

T1348-500G T1348-25G
TCT1348-500GEA 435 USD InStock
TCT1348-500G TCT1348-25G
(±)-Tetrahydro-2-furoic acid ≥98.0%
(±)-Tetrahydro-2-furoic acid
CAS Number: 16874-33-2
MDL Number: MFCD00022473
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 130
Flash Point (°C): 139
Specific Gravity (20/20): 1.21

Other Product Names / Synonyms: 2-Tetrahydrofuroic Acid


Formula: C₅H₈O₃
Boiling Pt: 130 °C
Density: 1.21 (20 °C)
Flash Pt: 139 °C
MDL Number: MFCD00022473
CAS Number: 16874-33-2
UN: 3265
ADR: 8,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f7ea1ed7335c9371963e75f6a6e63ccf7a88abe&vid=6f7ea1ed7335c9371963e75f6a6e63ccf7a88abe&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty