Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethoxytrimethylsilane ≥98.0%

Supplier: Decon Laboratories Ltd

T1394-250ML T1394-25ML
TCT1394-250MLEA 145 USD InStock
TCT1394-250ML TCT1394-25ML
Ethoxytrimethylsilane ≥98.0%
Ethoxytrimethylsilane
CAS Number: 1825-62-3
MDL Number: MFCD00009069
Molecular Formula: C5H14OSi
Molecular Weight: 118.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 75
Melting point (°C): -83
Flash Point (°C): -18
Specific Gravity (20/20): 0.76

Other Product Names / Synonyms: Ethoxytrimethylsilane


Formula: C₅H₁₄OSi
Boiling Pt: 75 °C
Melting Pt: -83 °C
Density: 0.76 (20 °C)
Flash Pt: -18 °C
MDL Number: MFCD00009069
CAS Number: 1825-62-3
EINECS: 217-370-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51728be01ee1bdf191b77338f2bb080ec2d0d749&vid=51728be01ee1bdf191b77338f2bb080ec2d0d749&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty