Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Valeric acid ≥98.0%

Supplier: Decon Laboratories Ltd
Danger

V0003-25ML V0003-500ML
TCV0003-25MLEA 23.4 USD InStock
TCV0003-25ML TCV0003-500ML
Valeric acid ≥98.0%
Valeric acid
CAS Number: 109-52-4
MDL Number: MFCD00004413
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 186
Melting point (°C): -18
Flash Point (°C): 96
Specific Gravity (20/20): 0.94

Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Lipids
Other Product Names / Synonyms: Pentanoic Acid


Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 186
Melting Pt: -32
Density: 0.94 (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00004413
CAS Number: 109-52-4
EINECS: 203-677-2
UN: 3265
ADR: 8 III
Merck Index: 12,10042
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11c5107eb93cde517daee7119af10553509d0e7c&vid=11c5107eb93cde517daee7119af10553509d0e7c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty