Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl valerate ≥98.0%

Supplier: Decon Laboratories Ltd

V0004-25ML V0004-500ML
TCV0004-025MLEA 21.96 USD InStock
TCV0004-025ML TCV0004-500ML
Ethyl valerate ≥98.0%
Ethyl valerate
CAS Number: 539-82-2
MDL Number: MFCD00009479
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 145
Flash Point (°C): 41
Specific Gravity (20/20): 0.88

Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Valeric Acid Ethyl Ester


Formula: C₇H₁₄O₂
Boiling Pt: 145 °C
Melting Pt: -91 °C
Density: 0.88 (20 °C)
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00009479
CAS Number: 539-82-2
EINECS: 208-726-1
UN: 3272
ADR: 3,III
Merck Index: 12,10042
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty