Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3-methoxybenzyl alcohol ≥98.0%

Supplier: Decon Laboratories Ltd

V0018-500G V0018-25G
TCV0018-500GEA 612 USD InStock
TCV0018-500G TCV0018-025G
4-Hydroxy-3-methoxybenzyl alcohol ≥98.0%
4-Hydroxy-3-methoxybenzyl alcohol
CAS Number: 498-00-0
MDL Number: MFCD00004659
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 114

Other Product Names / Synonyms: 4-Hydroxy-3-methoxybenzyl Alcohol


Formula: C₈H₁₀O₃
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 114 °C
MDL Number: MFCD00004659
CAS Number: 498-00-0
EINECS: 207-852-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dfec7d061e80aa047c4ca12d451b29622c5dd15&vid=5dfec7d061e80aa047c4ca12d451b29622c5dd15&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty