Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(cyclopentadienyl)zirconium dichloride ≥97.0%

Supplier: Decon Laboratories Ltd

Z0007-25G Z0007-5G
TCZ0007-25GEA 164.74 USD InStock
TCZ0007-25G TCZ0007-5G
Bis(cyclopentadienyl)zirconium dichloride ≥97.0%
Bis(cyclopentadienyl)zirconium dichloride
CAS Number: 1291-32-3
MDL Number: MFCD00003726
Molecular Formula: C10H10Cl2Zr
Molecular Weight: 292.31
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White

Product Categories: Synthetic Organic Chemistry, Glycosylation, Materials Chemistry, Polymerization Catalysts, Analytical Science, Chemical Shift Standards for Multinuclear NMR
Other Product Names / Synonyms: Bis(cyclopentadienyl)zirconium Dichloride


Formula: C₁₀H₁₀Cl₂Zr
MDL Number: MFCD00003726
CAS Number: 1291-32-3
EINECS: 215-066-8
UN: 3261
ADR: 8,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41a8328928ab2ad4bbc3f3417fcaa904f6af969f&vid=41a8328928ab2ad4bbc3f3417fcaa904f6af969f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty