Cole-Parmer® Essentials Sterile Erlenmeyer Flasks with Cap, PETG
Supplier: Argos Technologies
34522-40 34522-41 34522-42 34522-43 34522-44 34522-45
77107-074CS 115.48 USD InStock
77107-074 77107-082 77107-076 77107-084 77107-078 77107-086
Cole-Parmer® Essentials Sterile Erlenmeyer Flasks with Cap, PETG
Flasks Erlenmeyer Flasks
Sterile Erlenmeyer flasks ideal for media preparation, mixing cell cultures, and storage.Sterile Erlenmeyer flasks ideal for media preparation, mixing cell cultures, and storage.
  • Gamma sterilized (SAL 10-6)
  • DNase/RNase free and nonpyrogenic
  • Withstands temperature range of –60 to 60 °C (–76 to 140 °F)
  • Liquid sticking–resistant neck for easier pouring
  • Includes HDPE cap; choice of plug seal or vented
Sterile polyethylene terephthalate glycol (PETG) Erlenmeyer flasks are ideal for containing, mixing, and storing solutions and are a safe and shatterproof alternative to glass. These crystal-clear flasks feature molded-in graduations allowing you quick volume assessment. Liquid sticking-resistant design at the neck repels liquid for easier pouring. They have a temperature range of –60 to 60 °C (–76 to 140 °F) and are not autoclavable.
Certificaciones:
Manufactured in an ISO Class 8 (formerly Class 100,000) cleanroom environment, these individually packaged Erlenmeyer flasks are sterilized by gamma irradiation per ISO 11137-2 (SAL 10-6), and are ISO 13485 and ISO 9001 certified, DNase/RNase free, and nonpyrogenic.
Información para pedidos:
HDPE caps are included. Choose from a leakproof plug seal cap or a vented cap with 0.22 µm hydrophobic PTFE membrane for continuous gas exchange.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty