XRF Thin Film
XRF Thin Film
Precision-crafted of stainless steel, with a highly polished finish. Weights 30kg to 5kg are flat on top with a recessed grip handle for easy handling and stacking. Weights 5kg to 1kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping. Weights below 1kg are straight cylinders. All are manufactured with adjusting cavities.
Weights 5kg and below supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove.
VWR offers a wide assortment of products and brands in this category. If you can not find the product or brand that you are looking for in this listing, please use the general search field in the top-left corner of the screen to search the entire VWR portfolio.