Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hamster IgG Isotype Control (FITC (Fluorescein Isothiocyanate))

Supplier: Southern Biotechnology
0121-02
100242-136EA 167.38 USD InStock
100242-136
Hamster IgG Isotype Control (FITC (Fluorescein Isothiocyanate))
Antibodies
Hamster IgG-FITC for use as a control in flow cytometry assays.

Type: Control
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty