Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD8 alpha Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 76-2-11]

Supplier: Southern Biotechnology
4520-09
100244-858EA 325.62 USD InStock
100244-858
Anti-CD8 alpha Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 76-2-11]
Antibodies
Porcine CD8, a type I transmembrane glycoprotein, is expressed as either a CD8αα homodimer or a CD8αβ heterodimer. It is found on most thymocytes and on the suppressor/cytotoxic subpopulation of peripheral T cells. CD8 functions primarily as a co-receptor with MHC Class I-restricted T cell receptors in antigen recognition. The monoclonal antibody 76-2-11 reacts with the CD8 α-chain.

Type: Primary
Antigen: CD8alpha
Clonality: Monoclonal
Clone: 76-2-11
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Porcine

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty