Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgG (H+L) Rabbit Antibody (AP (Alkaline Phosphatase))

Supplier: Southern Biotechnology
6135-04
100245-094EA 281.15 USD InStock
100245-094
Anti-IgG (H+L) Rabbit Antibody (AP (Alkaline Phosphatase))
Antibodies
Rabbit F(ab')2 Anti-Rat IgG(H+L)-AP antibody with minimal reactivity to human proteins for use in ELISA and immunocytochemistry assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty