Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgA Mouse Monoclonal Antibody (Biotin) [clone: A-1]

Supplier: Southern Biotechnology
8330-08
100245-530EA 461.62 USD InStock
100245-530
Anti-IgA Mouse Monoclonal Antibody (Biotin) [clone: A-1]
Antibodies
Biotin Anti-Chicken IgA antibody for use in flow cytometry, immunohistochemistry / immunocytochemistry, ELISA, and immunoprecipitation assays.

Type: Primary
Antigen: IgA
Clonality: Monoclonal
Clone: A-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Chicken

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty