Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD57 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: NK-1]

Supplier: Southern Biotechnology
9665-09
100246-340EA 341.31 USD InStock
100246-340
Anti-CD57 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: NK-1]
Antibodies
CD57 represents a 110 kDa oligosaccharide antigenic determinant present on a variety of polypeptides, lipids, and chondroitan sulfate proteoglycans. It is expressed on 7-35% of normal peripheral blood lymphocytes, including subsets of NK cells and CD8+ T lymphocytes. The carbohydrate is expressed on various cell adhesion molecules in the nervous system and is suggested to play a role in cell–cell and cell–substrate interactions.

Type: Primary
Antigen: CD57
Clonality: Monoclonal
Clone: NK-1
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty