Anti-CD107a Mouse Monoclonal Antibody [clone: H4A3]
Supplier: Southern Biotechnology
9835-01
100246-514EA 142.54 USD InStock
100246-514
Anti-CD107a Mouse Monoclonal Antibody [clone: H4A3]
Antibodies
CD107a, also known as lysosomal-associated membrane protein 1 (LAMP-1), is a heavily glycosylated type I transmembrane protein that constitutes the major sialoglycoproteins on lysosomal membranes. It is a ligand for galaptin, an S-type lectin present in extracellular matrix, through its recognition of acetyllactosamine oligosaccharide chains, and is a ligand for E-selectin-mediated cell adhesion. CD107a is expressed by activated T cells, macrophages, dendritic cells, activated platelets, tonsillar epithelium, and some tumor cell lines, including U937 and KG1a. It is also a widely expressedMore Product Information
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty