Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD42b Mouse Monoclonal Antibody (Spectral Red® RPE (R-Phycoerythrin)/Cy5®) [clone: MM2/174]

Supplier: Southern Biotechnology
9615-13
101442-970EA 447.23 USD InStock
101442-970
Anti-CD42b Mouse Monoclonal Antibody (Spectral Red® RPE (R-Phycoerythrin)/Cy5®) [clone: MM2/174]
Antibodies
CD42b, also known as GPIbα, is a two-chain membrane glycoprotein that forms a noncovalent complex with CD42a (GPIX) on the surface of platelets and megakaryocytes. Binding of the GPIb/GPIX heterodimer to von Willebrand factor bound to exposed subendothelial surfaces is essential for platelet adhesion at sites of injury.

Type: Primary
Antigen: CD42b
Clonality: Monoclonal
Clone: MM2/174
Conjugation: Spectral Red® RPE (R-Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty