Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IgG2a Goat Antibody

Supplier: Southern Biotechnology
1083-01
103259-792EA 272 USD InStock
103259-792
Anti-IgG2a Goat Antibody
Antibodies
Purified Goat Anti-Mouse IgG2a antibody with minimal reactivity to human, bovine, and horse serum proteins for use in ELISA assays.

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty